Abnova
Manufacturer Code:H00009056K
Catalog # 89-203-012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC7A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC7A7 peptide using ARM Technology.
For Research Use Only
Protein Aliases: LAT3 LPI MOP-2 Y+LAT1 y+LAT-1; Monocyte amino acid permease 2; MOP-2; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; Solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+LAT1
Gene Aliases: LAT3; LPI; MOP-2; SLC7A7; y+LAT-1; Y+LAT1
UniProt ID: (Human) Q9UM01
Entrez Gene ID: (Human) 9056
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.