Invitrogen
Manufacturer Code:PA571365
Catalog # PIPA571365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of mouse SLC7A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4F2 LC; 4F2 light chain; L-type amino acid transporter 1; Large neutral amino acids transporter small subunit 1; LAT1; SLC7A5 solute carrier family 7 (cationic amino acid transporter y+ system) member 5 4F2LC CD98 D16S469E E16 LAT1 MPE16 hLAT1 4F2 light chain CD98 light chain L-type amino acid transporter 1 integral membrane protein E16 large neutral amino acids transporter 1 large neutral amino acids transporter small subunit 1 sodium-independent neutral amino acid transporter LAT1; Solute carrier family 7 member 5
Gene Aliases: 4F2LC; D0H16S474E; LAT1; Slc7a5; TA1
UniProt ID: (Mouse) Q9Z127
Entrez Gene ID: (Mouse) 20539
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.