Invitrogen
Manufacturer Code:PA5115916
Catalog # PIPA5115916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC7A5 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4F2 4F2 LC 4F2 light chain 4F2HC 4F2LC 4T2HC CD98 CD98 light chain CD98HC CD98LC D0H16S474E D16S469E E16 hLAT1 Integral membrane protein E16 large neutral amino acids transporter 1 large neutral amino acids transporter small subunit 1 Lat1 LAT-1 L-type amino acid transporter 1 MDU1 MP hLAT1 MPE16 NACAE Protein TA1 SLC7A5 sodium-independent neutral amino acid transporter LAT1 solute carrier family 7 (amino acid transporter light chain L system) member 5 solute carrier family 7 (cationic amino acid transporter y+ system) member 5 solute carrier family 7 member 5 TA1 tumor-associated protein 1 y+ system cationic amino acid transporter; 4F2 LC; 4F2 light chain; CD98 light chain; E16; hLAT1; Integral membrane protein E16; L-type amino acid transporter 1; large neutral amino acids transporter 1; Large neutral amino acids transporter small subunit 1; LAT1; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; Solute carrier family 7 member 5; y+ system cationic amino acid transporter
Gene Aliases: 4F2LC; CD98; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; LAT1; MPE16; SLC7A5; TA1
UniProt ID: (Human) Q01650, (Mouse) Q9Z127
Entrez Gene ID: (Human) 8140, (Mouse) 20539
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.