Proteintech
Manufacturer Code:137521AP150UL
Catalog # 50-555-715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC7A5 Fusion Protein Ag4694 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4F2 LC; 4F2 light chain; 4F2 light chain 4F2LC CD98 light chain CD98LC D16S469E E16 hLAT1 Integral membrane protein E16 LAT1 MPE16 SLC7A5; 4F2LC; CD98 light chain; E16; hLAT1; Integral membrane protein E16; L-type amino acid transporter 1; large neutral amino acids transporter 1; Large neutral amino acids transporter small subunit 1; LAT-1; LAT1; Protein TA1; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; Solute carrier family 7 member 5; tumor-associated protein 1; y+ system cationic amino acid transporter
Gene Aliases: 4F2LC; CD98; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; LAT1; MPE16; SLC7A5; TA1
UniProt ID: (Human) Q01650, (Mouse) Q9Z127, (Rat) Q63016
Entrez Gene ID: (Human) 8140, (Mouse) 20539, (Rat) 50719
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.