Invitrogen
Manufacturer Code:PA5101500
Catalog # PIPA5101500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC7A4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAT-4; CAT4 CAT-4 Cationic amino acid transporter 4 HCAT3 Ig heavy chain variable region SLC7A4 solute carrier family 7 (cationic amino acid transporter y+ system) member 4 solute carrier family 7 (orphan transporter) member 4 solute carrier family 7 member 4 solute carrier family 7 member 4 VH VH 3 family; Cationic amino acid transporter 4; Ig heavy chain variable region; solute carrier family 7 (cationic amino acid transporter, y+ system), member 4; solute carrier family 7 (orphan transporter), member 4; Solute carrier family 7 member 4; solute carrier family 7, member 4; VH 3 family
Gene Aliases: CAT-4; CAT4; HCAT3; SLC7A4; VH
UniProt ID: (Human) O43246
Entrez Gene ID: (Human) 6545, (Rat) 303787
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.