Abnova
Manufacturer Code:PAB22772
Catalog # 89-124-478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC7A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC7A4.
For Research Use Only
Protein Aliases: CAT-4; CAT4; Cationic amino acid transporter 4; HCAT3; Ig heavy chain variable region; MGC129976; MGC129977; solute carrier family 7 (cationic amino acid transporter, y+ system), member 4; solute carrier family 7 (orphan transporter), member 4; Solute carrier family 7 member 4; solute carrier family 7, member 4; VH; VH 3 family
Gene Aliases: CAT-4; CAT4; HCAT3; SLC7A4; VH
UniProt ID: (Human) O43246
Entrez Gene ID: (Human) 6545
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.