Invitrogen
Manufacturer Code:PA569257
Catalog # PIPA569257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SLC7A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter, cationic 3; ATRC3 CAT3; CAT-3; Cationic amino acid transporter 3; Cationic amino acid transporter y+; solute carrier family 7 (cationic amino acid transporter, y+ system), member 3; Solute carrier family 7 member 3; solute carrier family 7, member 3
Gene Aliases: ATRC3; CAT-3; CAT3; SLC7A1; SLC7A2; SLC7A3
UniProt ID: (Human) Q8WY07, (Mouse) P70423, (Rat) O08812
Entrez Gene ID: (Bovine) 539065, (Rabbit) 100347183, (Human) 84889, (Horse) 100055782, (Dog) 491945, (Guinea pig) 100715043, (Mouse) 11989, (Rat) 29485
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.