Invitrogen
Manufacturer Code:PA569252
Catalog # PIPA569252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC7A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20.5; amino acid transporter, cationic 2 (low affinity); ATRC2 CAT2 HCAT2 low affinity cationic amino acid transporter 2; CAT-2; Cationic amino acid transporter 2; cationic amino acid transporter 2A; cationic amino acid transporter 2B; cationic amino acid transporter-2A; Low affinity cationic amino acid transporter 2; solute carrier family 7 (cationic amino acid transporter, y+ system), member 2; Solute carrier family 7 member 2; solute carrier family 7, member 2; T-cell early activation protein; TEA
Gene Aliases: 20.5; AI158848; ATRC2; CAT-2; CAT2; Cat2a; Cat2b; HCAT2; SLC7A2; Tea
UniProt ID: (Human) P52569, (Pig) A8I499, (Mouse) P18581, (Rat) B5D5N9
Entrez Gene ID: (Human) 6542, (Dog) 475614, (Pig) 100037298, (Horse) 100050240, (Rabbit) 100346689, (Mouse) 11988, (Rat) 64554
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.