Invitrogen
Manufacturer Code:PA5116134
Catalog # PIPA5116134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human xCT corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9930009M05Rik AI451155 Amino acid transport system xc- calcium channel blocker resistance protein CCBR1 CCBR1 cysteine/glutamate transporter Cystine/glutamate transporter Slc7a11 sodium independent anionic amino acid transport system solute carrier family 7 (anionic amino acid transporter light chain xc- system) member 11 solute carrier family 7 (cationic amino acid transporter y+ system) member 11 solute carrier family 7 member 11 solute carrier family 7 (cationic amino acid transporter y+ system) member 11 sut xCT; Amino acid transport system xc-; Calcium channel blocker resistance protein CCBR1; cysteine/glutamate transporter; Cystine/glutamate transporter; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; Solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; xCT
Gene Aliases: 9930009M05Rik; AI451155; CCBR1; SLC7A11; sut; xCT
UniProt ID: (Human) Q9UPY5, (Mouse) Q9WTR6
Entrez Gene ID: (Human) 23657, (Mouse) 26570
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.