Invitrogen
Manufacturer Code:PA5114947
Catalog # PIPA5114947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CAT1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4831426K01Rik AI447493 amino acid transporter cationic 1 Atrc1 Atrc-1 CAT1 CAT-1 cationic amino acid transporter 1 cationic amino acid transporter y+ system cationinc amino acid transporter 1 ecotropic retroviral leukemia receptor Ecotropic retroviral leukemia receptor homolog ecotropic retroviral receptor ecotropic retrovirus receptor Ecotropic retrovirus receptor homolog ERR HCAT1 high affinity cationic amino acid transporter 1 mCAT-1 Rec-1 REC1L Rev-1 Slc7a1 solute carrier family 7 (cationic amino acid transporter y+ system) member 1 solute carrier family 7 member 1 Solute carrier family 7 member A1 (amino acid transporter cationic 1) solute carrier family 7 member 1 system Y+ basic amino acid transporter; amino acid transporter, cationic 1; CAT-1; CAT1; cationic amino acid transporter 1; cationic amino acid transporter, y+ system; cationinc amino acid transporter 1; Ecotropic retroviral leukemia receptor; Ecotropic retroviral leukemia receptor homolog; ecotropic retroviral receptor; Ecotropic retrovirus receptor; Ecotropic retrovirus receptor homolog; ERR; High affinity cationic amino acid transporter 1; solute carrier family 7 (cationic amino acid transporter, y+ system), member 1; Solute carrier family 7 member 1; Solute carrier family 7 member A1 (amino acid transporter cationic 1); solute carrier family 7, member 1; System Y+ basic amino acid transporter
Gene Aliases: 4831426K01Rik; AI447493; Atrc-1; ATRC1; CAT-1; CAT1; ERR; HCAT1; mCAT-1; Rec-1; REC1L; Rev-1; SLC7A1
UniProt ID: (Human) P30825, (Mouse) Q09143
Entrez Gene ID: (Human) 6541, (Mouse) 11987, (Rat) 25648
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.