Invitrogen
Manufacturer Code:PA542132
Catalog # PIPA542132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC6A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: choline transporter; CHOT1; creatine transporter 1; creatine transporter SLC6A8 variant D; creatine transporter, solute carrier family 6, member 8; CT1; Na+ dependent transporter; Sodium- and chloride-dependent creatine transporter 1; Solute carrier family 6 (neurotransmitter transporter creatine) member 8 CRTR CT1 MGC87396 CRT; solute carrier family 6 (neurotransmitter transporter), member 8; solute carrier family 6 (neurotransmitter transporter, creatine), member 8; Solute carrier family 6 member 8; solute carrier family 6, member 8
Gene Aliases: AA589632; CCDS1; CHOT1; CHT1; Creat; CRT; CRTR; CT1; CTR5; SLC6A8
UniProt ID: (Bovine) O18875, (Human) P48029, (Rabbit) P31661, (Rat) P28570, (Mouse) Q8VBW1
Entrez Gene ID: (Bovine) 282367, (Dog) 492242, (Horse) 100058799, (Human) 6535, (Rabbit) 100009280, (Rat) 50690, (Guinea pig) 100728288, (Mouse) 102857
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.