Invitrogen
Manufacturer Code:PA577458
Catalog # PIPA577458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KPLMVHMRKYGGIT corresponding to amino acid residues 602- 615 of rat Proline Transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain-specific L-proline transporter; L-proline transporter; proline transporter; Proline Transporter (PROT); slc6a7; Sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; Solute carrier family 6 member 7
Gene Aliases: AW455934; PROT; SLC6A7
UniProt ID: (Human) Q99884, (Rat) P28573, (Mouse) Q6PGE7
Entrez Gene ID: (Human) 6534, (Rat) 117100, (Mouse) 240332
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.