Novus Biologicals
Manufacturer Code:NBP187216
Catalog # NBP187216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SLTSDLPWEHCGNWWNTELCLEHRVSKDGNGALPLNLTCTVSPSEEYWSRYVLHIQGSQGIGSPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain-specific L-proline transporter; brain-specific L-proline transporter PROTSolute carrier family 6 member 7 sodium-dependent proline transporter solute carrier family 6 (neurotransmitter transporter L-proline) member 7; Sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; Solute carrier family 6 member 7
Gene Aliases: PROT; SLC6A7
UniProt ID: (Human) Q99884
Entrez Gene ID: (Human) 6534
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.