Abnova
Manufacturer Code:PAB22226
Catalog # 89-123-931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC6A7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC6A7.
For Research Use Only
Protein Aliases: brain-specific L-proline transporter; PROT; Sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; Solute carrier family 6 member 7
Gene Aliases: PROT; SLC6A7
UniProt ID: (Human) Q99884
Entrez Gene ID: (Human) 6534
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.