Invitrogen
Manufacturer Code:PA518374
Catalog # PIPA518374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (LHQGERETWGK) corresponding to the internal amino acids of SLC6A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-hydroxytryptamine (serotonin) transporter; 5HT transporter; 5HTT; Na+/Cl- dependent serotonin transporter; serotonin transporter; serotonin transporter 1; SERT; SLC6A4 SERT solute carrier family 6 (neurotransmitter transporter serotonin) member 4 HGNC:11050 5-HTT HTT OCD1 hSERT 5-hydroxytryptamine transporter 5HT transporter Na+/Cl- dependent serotonin transporter serotonin transporter sodium-dependent serotonin transporter solute carrier family 6 member 4 5-HTTLPR OTTHUMP00000163633; Sodium-dependent serotonin transporter; solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; Solute carrier family 6 member 4; solute carrier family 6, member 4
Gene Aliases: 5-HTT; 5-HTTLPR; 5HTT; hSERT; HTT; OCD1; SERT; SERT1; SLC6A4
UniProt ID: (Human) P31645, (Rat) P31652
Entrez Gene ID: (Human) 6532, (Rat) 25553
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.