R&D Systems
Manufacturer Code:PPS069
Catalog # PPS069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC6A3/DAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long-term storage < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DA transporter; DA transporter DAT1 DATDAT1Solute carrier family 6 member 3 SLC6A3 sodium-dependent dopamine transporter solute carrier family 6 (neurotransmitter transporter dopamine) member 3 VNTR; dopamine transporter 1; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; Solute carrier family 6 member 3
Gene Aliases: DAT; DAT1; PKDYS; SLC6A3
UniProt ID: (Human) Q01959
Entrez Gene ID: (Human) 6531
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.