Novus Biologicals
Manufacturer Code:NBP128665
Catalog # NBP128665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NET-05 |
Immunogen | Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: NAT1neurotransmitter transporter NET NET1sodium-dependent noradrenaline transporter Norepinephrine transporter SLC6A5 solute carrier family 6 (neurotransmitter transporter noradrenalin) member 2 Solute carrier family 6 member 2 solute carrier family 6 member 5; NET; neurotransmitter transporter; Norepinephrine transporter; Sodium-dependent noradrenaline transporter; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; Solute carrier family 6 member 2; solute carrier family 6 member 5
Gene Aliases: NAT1; NET; NET1; SLC6A2; SLC6A5
UniProt ID: (Human) P23975
Entrez Gene ID: (Human) 6530
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.