Invitrogen
Manufacturer Code:PA568261
Catalog # PIPA568261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SLC6A17. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: orphan sodium- and chloride-dependent neurotransmitter transporter NTT4; Sodium-dependent neurotransmitter transporter NTT4; Sodium-dependent neutral amino acid transporter SLC6A17; Sodium-dependent neutral amino acid transporter SLC6A17 Sodium-dependent neurotransmitter transporter NTT4 Solute carrier family 6 member 17 SLC6A17 NTT4; sodium-dependent orphan neurotramsmitter transporter NTT4; solute carrier family 6 (neurotransmitter transporter), member 17; solute carrier family 6 (neutral amino acid transporter), member 17; Solute carrier family 6 member 17; solute carrier family 6, member 17
Gene Aliases: D130012J15Rik; MRT48; NTT4; Rxt1; SLC6A17
UniProt ID: (Human) Q9H1V8, (Rat) P31662, (Mouse) Q8C057
Entrez Gene ID: (Human) 388662, (Rat) 613226, (Mouse) 229706
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.