Abnova
Manufacturer Code:H00388662K
Catalog # 89-209-160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC6A17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC6A17 peptide using ARM Technology.
For Research Use Only
Protein Aliases: NTT4; orphan sodium- and chloride-dependent neurotransmitter transporter NTT4; Sodium-dependent neurotransmitter transporter NTT4; Sodium-dependent neutral amino acid transporter SLC6A17; solute carrier family 6 (neurotransmitter transporter), member 17; solute carrier family 6 (neutral amino acid transporter), member 17; Solute carrier family 6 member 17; solute carrier family 6, member 17
Gene Aliases: MRT48; NTT4; SLC6A17
UniProt ID: (Human) Q9H1V8
Entrez Gene ID: (Human) 388662
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.