Invitrogen
Manufacturer Code:PA587898
Catalog # PIPA587898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 350-450 of human SLC6A15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B0AT2 protein; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; Sodium- and chloride-dependent neurotransmitter transporter NTT73; Sodium-dependent neutral amino acid transporter B(0)AT2; Solute carrier family 6 member 15; Transporter v7-3
Gene Aliases: AA536730; AI326450; AI326451; B0at2; Ntt73; Slc6a15; v7-3
UniProt ID: (Mouse) Q8BG16
Entrez Gene ID: (Mouse) 103098
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.