Novus Biologicals
Manufacturer Code:NBP159366
Catalog # NBP159366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC6A15(solute carrier family 6 member 15) The peptide sequence was selected from the N terminal of SLC6A15. Peptide sequence DQCPLVKNASHTFVEPECEQSSATTYYWYREALNISSSISESGGLNWKMT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B0AT2 DKFZp761I0921 FLJ10316 homolog of rat orphan transporter v7-3 hv7-3 MGC87066 NTT73orphan sodium- and chloride-dependent neurotransmitter transporter NTT73 orphan transporter v7-3 SBAT1 Sodium- and chloride-dependent neurotransmitter transporter NTT73 sodium/chloride dependent neurotransmitter transporter Homo sapiens orphanneurotransmitter transporter NTT7 Sodium-coupled branched-chain amino-acid transporter 1 solute carrier family 6 (neurotransmitter transporter) member 15 solute carrier family 6 (neutral amino acid transporter) member 15 Solute carrier family 6 member 15 solute carrier family 6 member 15 Transporter v7-3 V7-3; homolog of rat orphan transporter v7-3; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; Sodium- and chloride-dependent neurotransmitter transporter NTT73; Sodium-coupled branched-chain amino-acid transporter 1; Sodium-dependent neutral amino acid transporter B(0)AT2; sodium/chloride dependent neurotransmitter transporter Homo sapiens orphan neurotransmitter transporter NTT7; solute carrier family 6 (neurotransmitter transporter), member 15; solute carrier family 6 (neutral amino acid transporter), member 15; Solute carrier family 6 member 15; solute carrier family 6, member 15; Transporter v7-3
Gene Aliases: B0AT2; hv7-3; NTT73; SBAT1; SLC6A15; V7-3
UniProt ID: (Human) Q9H2J7
Entrez Gene ID: (Human) 55117
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.