Invitrogen
Manufacturer Code:PA524005
Catalog # PIPA524005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 9-38 amino acids from the N-terminal region of human S6A12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: betaine/GABA transporter-1; BGT-1; BGT-1 BGT1 GAT2 Na(+)/Cl(-) betaine/GABA transporter gamma-aminobutyric acid transporter sodium- and chloride-dependent betaine transporter; gamma-aminobutyric acid transporter; Na(+)/Cl(-) betaine/GABA transporter; Sodium- and chloride-dependent betaine transporter; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; Solute carrier family 6 member 12
Gene Aliases: BGT-1; BGT1; GAT2; SLC6A12
UniProt ID: (Human) P48065
Entrez Gene ID: (Human) 6539
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.