Invitrogen
Manufacturer Code:PA542425
Catalog # PIPA542425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC6A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA transporter 1; GABA transporter GAT-1 homolog; GABA transporter GAT-1 homolog, similar to rat GABA transporter GAT-1, Swiss-Prot Accession Number P23978; GABA transporter protein; gamma-aminobutyric acid (GABA-A) transporter 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; Solute carrier family 6 (neurotransmitter transporter GABA) member 1 GABATHG GABATR GAT1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: A730043E01; GABATHG; GABATR; Gabt; GABT1; GAT-1; GAT1; MAE; SLC6A1; XT-1; Xtrp1
UniProt ID: (Human) P30531, (Rat) P23978, (Mouse) P31648
Entrez Gene ID: (Horse) 100051613, (Human) 6529, (Bovine) 407118, (Dog) 484655, (Rabbit) 103349186, (Rat) 79212, (Mouse) 232333
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.