Invitrogen
Manufacturer Code:PA533048
Catalog # PIPA533048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from C-terminus of human SLC5A9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSGLT4; Na(+)/glucose cotransporter 4; SGLT4 Na(+)/glucose cotransporter 4 hSGLT4 sodium/glucose cotransporter 4 solute carrier family 5 member 9; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; Solute carrier family 5 member 9
Gene Aliases: SGLT4; SLC5A9
UniProt ID: (Human) Q5TET3
Entrez Gene ID: (Human) 200010
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.