Abnova
Manufacturer Code:PAB28332
Catalog # 89-129-834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at internal region of human SLC5A9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyconal antibody raised against synthetic peptide of SLC5A9.
For Research Use Only
Protein Aliases: hSGLT4; MGC132517; MGC132523; Na(+)/glucose cotransporter 4; SGLT4; Sodium/glucose cotransporter 4; solute carrier family 5 (sodium/glucose cotransporter), member 9; solute carrier family 5 (sodium/sugar cotransporter), member 9; Solute carrier family 5 member 9
Gene Aliases: SGLT4; SLC5A9
UniProt ID: (Human) Q5TET3
Entrez Gene ID: (Human) 200010
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.