Invitrogen
Manufacturer Code:PA577385
Catalog # PIPA577385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DWNQTAYGYPDPK corresponding to amino acid residues 299-311 of mouse Choline Transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHT; Hemicholinium-3-sensitive choline transporter; High affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; Solute carrier family 5 member 7; solute carrier family 5, member 7
Gene Aliases: CHT; CHT1; HMN7A; SLC5A7
UniProt ID: (Human) Q9GZV3, (Mouse) Q8BGY9, (Rat) Q9JMD7
Entrez Gene ID: (Human) 60482, (Mouse) 63993, (Rat) 85426
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.