Invitrogen
Manufacturer Code:PA5117124
Catalog # PIPA5117124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 421-580 of human SLC5A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHT; CHT CHT1 hCHT Hemicholinium-3-sensitive choline transporter high affinity choline transporter 1 high affinity choline transporter hemicholinium-3-sensitive choline transporter HMN7A MGC126299 MGC126300 SLC5A7 solute carrier family 5 (choline transporter) member 7 solute carrier family 5 (sodium/choline cotransporter) member 7 solute carrier family 5 member 7 solute carrier family 5 member 7; Hemicholinium-3-sensitive choline transporter; High affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; Solute carrier family 5 member 7; solute carrier family 5, member 7
Gene Aliases: CHT; CHT1; HMN7A; SLC5A7
UniProt ID: (Human) Q9GZV3, (Mouse) Q8BGY9, (Rat) Q9JMD7
Entrez Gene ID: (Human) 60482, (Mouse) 63993, (Rat) 85426
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.