Proteintech
Manufacturer Code:264071AP150UL
Catalog # 50-173-5326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC5A6 Fusion Protein Ag23907 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; SLC5A6 SMVT; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; Solute carrier family 5 member 6; solute carrier family 5, member 6
Gene Aliases: E430023I20; SLC5A6; SMVT
UniProt ID: (Human) Q9Y289, (Mouse) Q5U4D8, (Rat) O70247
Entrez Gene ID: (Human) 8884, (Mouse) 330064, (Rat) 170551
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.