Invitrogen
Manufacturer Code:PA533044
Catalog # PIPA533044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from C-terminus of human SLC5A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DJ90G24.4 SAAT1 SGLT3 Na(+)/glucose cotransporter 3 low affinity sodium glucose cotransporter low affinity sodium-glucose cotransporter sodium transporter sodium/glucose cotransporter 3 solute carrier family 5 (neutral amino acid transporters system A) member 4 solute carrie; low affinity sodium glucose cotransporter; Na(+)/glucose cotransporter 3; sodium transporter; sodium/glucose cotransporter 3; solute carrier family 5 (glucose activated ion channel), member 4; solute carrier family 5 (low affinity glucose cotransporter), member 4; solute carrier family 5 (neutral amino acid transporters, system A), member 4; Solute carrier family 5 member 4
Gene Aliases: DJ90G24.4; SAAT1; SGLT3; SLC5A4
UniProt ID: (Human) Q9NY91
Entrez Gene ID: (Human) 6527
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.