Invitrogen
Manufacturer Code:PA533041
Catalog # PIPA533041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-terminus of human SLC5A3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/myo-inositol cotransporter; SMIT SMIT1 SMIT2 BCW2 protein Na(+)/myo-inositol cotransporter human solute carrier family 5 member 3 Sodium/myo-inositol cotransporter sodium/myo-inositol cotransporter sodium/myo-inositol cotransporter 1 sodium/myo-inositol transporter 1 solute carrier family 5 (inosi; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3
Gene Aliases: BCW2; SMIT; SMIT1; SMIT2
Entrez Gene ID: (Human) 6526
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.