Invitrogen
Manufacturer Code:PA5101896
Catalog # PIPA5101896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC5A3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA623876 BCW2 BCW2 protein BF642829 Na(+)/myo-inositol cotransporter Slc5a3 SMIT Smit1 SMIT2 sodium myo-inositol transporter 1 sodium/myo-inositol cotransporter sodium/myo-inositol cotransporter 1 Sodium/myo-inositol transporter 1 solute carrier family 5 (inositol transporters) member 3 solute carrier family 5 (sodium/myo-inositol cotransporter) member 3 Solute carrier family 5 member 3 solute carrier family 5 member 3 solute carrier family 5 member alpha 3; Na(+)/myo-inositol cotransporter; SMIT1; sodium myo-inositol transporter 1; Sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; Sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; Solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3
Gene Aliases: AA623876; BCW2; BF642829; Slc5a3; SMIT; SMIT1; SMIT2
UniProt ID: (Mouse) Q9JKZ2
Entrez Gene ID: (Human) 6526, (Mouse) 53881, (Rat) 114507
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.