Abnova
Manufacturer Code:H00006526A01
Catalog # 89-105-459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 533 a.a. &bsim 641 a.a) partial recombinant protein with GST tag.,SLC5A3 (NP_008864 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC5A3.
For Research Use Only
Protein Aliases: Na(+)/myo-inositol cotransporter; SMIT; SMIT2; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3
Gene Aliases: BCW2; SMIT; SMIT1; SMIT2
Entrez Gene ID: (Human) 6526
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.