Novus Biologicals
Manufacturer Code:NBP293974100UL
Catalog # NB022513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 200-280 of human SLC5A12 (NP_848593.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral sodium monocarboxylate cotransporter; Electroneutral sodium monocarboxylate cotransporter Low-affinity sodium-lactate cotransporter MGC52019 SMCT2DKFZp564G223 sodium-coupled monocarboxylate transporter 2 sodium-iodide related cotransporter solute carrier family 5 (sodium/glucose cotransporter) member 12 Solute carrier family 5 member 12; Low-affinity sodium-lactate cotransporter; Sodium-coupled monocarboxylate transporter 2; sodium-iodide related cotransporter; solute carrier family 5 (sodium/glucose cotransporter), member 12; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 12; Solute carrier family 5 member 12
Gene Aliases: SLC5A12; SMCT2
UniProt ID: (Human) Q1EHB4
Entrez Gene ID: (Human) 159963
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.