Invitrogen
Manufacturer Code:PA589042
Catalog # PIPA589042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 540-640 of human SLC5A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of rabbit KST1; Na(+)/myo-inositol cotransporter 2; na+/myo-inositol cotransporter 2; putative sodium-coupled cotransporter RKST1; rkST1; slc5a11; slc5a11 {ECO:0000312|RGD:621410}; SMIT2; sodium-cotransporter rkST1; sodium-coupled cotransporter KST1; Sodium-dependent glucose cotransporter; Sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; Sodium/myo-inositol transporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; Solute carrier family 5 member 11
Gene Aliases: 2010013B02Rik; KST1; RKST1; RKST2; SGLT6; Slc5a10; SLC5A11; SLGTX; SMIT2
UniProt ID: (Human) Q8WWX8, (Rat) Q9Z1F2, (Mouse) Q8K0E3
Entrez Gene ID: (Human) 115584, (Rat) 252854, (Mouse) 233836
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.