Invitrogen
Manufacturer Code:PA569282
Catalog # PIPA569282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC5A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of rabbit KST1; KST1 RKST1 SGLT6 SMIT2 Na(+)/myo-inositol cotransporter 2 homolog of rabbit KST1 putative sodium-coupled cotransporter RKST1 sodium-dependent glucose cotransporter sodium/glucose cotransporter KST1 sodium/myo-inositol cotransporter 2 sodium/myo-inositol transporter 2; Na(+)/myo-inositol cotransporter 2; na+/myo-inositol cotransporter 2; Na+/myo-inositol cotransporter SMIT2; putative sodium-coupled cotransporter RKST1; rKST1; SLC5A11; slc5a11 {ECO:0000312|RGD:621410}; SMIT2; sodium-cotransporter rkST1; sodium-coupled cotransporter KST1; Sodium-dependent glucose cotransporter; sodium-glucose cotransporter-like protein; Sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; Sodium/myo-inositol transporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 10; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; Solute carrier family 5 member 11
Gene Aliases: 2010013B02Rik; KST1; RKST1; RKST2; SGLT6; Slc5a10; SLC5A11; SLGTX; SMIT2; zgc:92080
UniProt ID: (Rabbit) Q28728, (Human) Q8WWX8, (Bovine) Q3ZC26, (Rat) Q9Z1F2, (Mouse) Q8K0E3
Entrez Gene ID: (Rabbit) 100008981, (Human) 115584, (Dog) 489965, (Bovine) 539084, (Horse) 100068437, (Goat) 100861027, (Sheep) 101107154, (Guinea pig) 100722853, (Rat) 252854, (Mouse) 233836, (Zebrafish) 492334
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.