Novus Biologicals
Manufacturer Code:NBP15963520UL
Catalog # NBP15963520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC5A11(solute carrier family 5 (sodium/glucose cotransporter) member 11) The peptide sequence was selected from the middle region of SLC5A11. Peptide sequence VGGMEGLKEKYFLALASNRSENSSCGLPREDAFHIFRDPLTS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of rabbit KST1; KST1RKST1 Na(+)/myo-inositol cotransporter 2 na+/myo-inositol cotransporter 2 putative sodium-coupled cotransporter RKST1 SGLT6 SLGTX SMIT2homolog of rabbit KST1 Sodium/glucose cotransporter KST1 sodium/myo-inositol cotransporter 2 Sodium-dependent glucose cotransporter solute carrier family 5 (sodium/glucose cotransporter) member 11 Solute carrier family 5 member 11; Na(+)/myo-inositol cotransporter 2; putative sodium-coupled cotransporter RKST1; SMIT2; Sodium-dependent glucose cotransporter; Sodium/glucose cotransporter KST1; Sodium/myo-inositol cotransporter 2; Sodium/myo-inositol transporter 2; solute carrier family 5 (sodium/glucose cotransporter), member 11; solute carrier family 5 (sodium/inositol cotransporter), member 11; Solute carrier family 5 member 11
Gene Aliases: KST1; RKST1; SGLT6; SLC5A11; SLGTX; SMIT2
UniProt ID: (Human) Q8WWX8
Entrez Gene ID: (Human) 115584
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.