Novus Biologicals
Manufacturer Code:NBP29390020UL
Catalog # NB029659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human SLC4A8 (NP_001035049.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761B2318 Electroneutral Na(+)-driven Cl-HCO3 exchanger FLJ46462 KIAA0739 k-NBC3 Na+-driven Cl-HCO3 exchanger NBC NBC3electroneutral sodium bicarbonate exchanger 1 NDCBE NDCBE1 Solute carrier family 4 member 8 solute carrier family 4 sodium bicarbonate cotransporter member 8; Electroneutral Na(+)-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; k-NBC3; Solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Aliases: KIAA0739; NBC; NBC3; NDCBE; NDCBE1; SLC4A8
UniProt ID: (Human) Q2Y0W8
Entrez Gene ID: (Human) 9498
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.