Proteintech
Manufacturer Code:125311AP150UL
Catalog # 50-172-8044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC4A8 Fusion Protein Ag3227 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral Na(+)-driven Cl-HCO3 exchanger; Electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; k NBC3 KIAA0739 NBC NBC3 NDCBE1 SLC4A8; k-NBC3; Na-driven Cl-HCO3 exchanger NDCBE1-A; Na-driven Cl-HCO3 exchanger NDCBE1-B; Na-driven Cl-HCO3 exchanger NDCBE1-C; slc4a8; slc4a8 {ECO:0000312|RGD:735164}; sodium bicarbonate cotransporter isoform 3 kNBC-3; solute carrier family 4 (anion exchanger), member 8; Solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Aliases: AW493845; C230026C11; KIAA0739; kNBC-3; NBC; NBC3; NDCBE; NDCBE1; SLC4A8
UniProt ID: (Human) Q2Y0W8, (Mouse) Q8JZR6, (Rat) Q6RVG2
Entrez Gene ID: (Human) 9498, (Mouse) 59033, (Rat) 315311
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.