Novus Biologicals
Manufacturer Code:NBP232020
Catalog # NBP232020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: KKKGSLDSDNDDSDCPYSEKVPSIKIPMDIMEQQPFLSDSKPSDRERSPTFLERHTS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electrogenic sodium bicarbonate cotransporter 1; electrogenic sodium bicarbonate cotransporter 1 hhNMC HNBC1 KNBC kNBC1 Na(+)/HCO3(-) cotransporter NBC NBC1DKFZp781H1314 NBC2 NBCE1 pNBC SLC4A5 Sodium bicarbonate cotransporter sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter kidneysodium bicarbonate cotransporter pancreas) Solute carrier family 4 member 4 solute carrier family 4 sodium bicarbonate cotransporter member 4 solute carrier family 4 sodium bicarbonate cotransporter member 4 brain type solute carrier family 4 sodium bicarbonate cotransporter member 5; kNBC1; Na(+)/HCO3(-) cotransporter; Sodium bicarbonate cotransporter; sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter, kidney; sodium bicarbonate cotransporter, pancreas); solute carrier family 4 (sodium bicarbonate cotransporter), member 4; Solute carrier family 4 member 4; solute carrier family 4, sodium bicarbonate cotransporter, member 4, brain type; solute carrier family 4, sodium bicarbonate cotransporter, member 5
Gene Aliases: hhNMC; HNBC1; KNBC; kNBC1; NBC; NBC1; NBC2; NBCE1; NBCe1-A; pNBC; SLC4A4; SLC4A5
UniProt ID: (Human) Q9Y6R1
Entrez Gene ID: (Human) 8671
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.