Novus Biologicals
Manufacturer Code:NBP29437220UL
Catalog # NB023298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SLC4A11 (NP_114423.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bicarbonate transporter related protein 1; Bicarbonate transporter-related protein 1; Bicarbonate transporter-related protein 1 BTR1bicarbonate transporter related protein 1 CDPD CHED2 corneal endothelial dystrophy 2 (autosomal recessive) dJ794I6.2 FECD4 MGC126418 MGC126419 NaBC1 sodium bicarbonate transporter-like protein 11 Sodium borate cotransporter 1 sodium-coupled borate cotransporter 1 Solute carrier family 4 member 11 solute carrier family 4 sodium bicarbonate transporter-like member 11 solute carrier family 4 sodium borate transporter member 11; NaBC1; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; Solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11
Gene Aliases: BTR1; CDPD1; CHED; CHED2; dJ794I6.2; NABC1; SLC4A11
UniProt ID: (Human) Q8NBS3
Entrez Gene ID: (Human) 83959
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.