Abnova
Manufacturer Code:PAB16074
Catalog # 89-118-041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human SLC4A11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC4A11.
For Research Use Only
Protein Aliases: bicarbonate transporter related protein 1; Bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; Solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11
Gene Aliases: BTR1; CDPD1; CHED; CHED2; dJ794I6.2; NABC1; SLC4A11
UniProt ID: (Human) Q8NBS3
Entrez Gene ID: (Human) 83959
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.