Invitrogen
Manufacturer Code:PA577555
Catalog # PIPA577555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RAGLKSTKELIPTPAD corresponding to amino acid residues 481-496 of mouse MATE2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H(+)/organic cation antiporter kidney-specific; MATE-2; MATE2; mMATE-2; Multidrug and toxin extrusion protein 2; Solute carrier family 47 member 2
Gene Aliases: 4933429E10Rik; Mate2; Slc47a2
UniProt ID: (Mouse) Q3V050
Entrez Gene ID: (Mouse) 380701
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.