Invitrogen
Manufacturer Code:PA537989
Catalog # PIPA537989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence SECHVDFFRTPEEAH from the internal region near the C-terminus of SLC47A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Kidney-specific H(+)/organic cation antiporter; MATE-2; Multidrug and toxin extrusion protein 2; solute carrier family 47 (multidrug and toxin extrusion), member 2; Solute carrier family 47 member 2; solute carrier family 47 member 2 FLJ31196 MATE2 MATE2-B MATE2-K MATE2K H+/organic cation antiporter hMATE-2 kidney-specific H +/organic cation antiporter multidrug and toxin extrusion 2 multidrug and toxin
Gene Aliases: MATE2; MATE2-B; MATE2-K; MATE2K; SLC47A2
UniProt ID: (Human) Q86VL8
Entrez Gene ID: (Human) 146802
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.