Invitrogen
Manufacturer Code:PA524659
Catalog # PIPA524659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 125-153 amino acids from the N-terminal region of human S47A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H(+)/organic cation antiporter kidney-specific; Kidney-specific H(+)/organic cation antiporter; MATE-2; MATE2 MATE2-B MATE2-K MATE2K H+/organic cation antiporter MATE-2 hMATE-2 kidney-specific H(+)/organic cation antiporter multidrug and toxin extrusion 2 multidrug and toxin extrusion protein 2; mMATE-2; Multidrug and toxin extrusion protein 2; solute carrier family 47 (multidrug and toxin extrusion), member 2; Solute carrier family 47 member 2
Gene Aliases: 4933429E10Rik; MATE2; MATE2-B; MATE2-K; MATE2K; SLC47A2
UniProt ID: (Human) Q86VL8, (Mouse) Q3V050
Entrez Gene ID: (Human) 146802, (Mouse) 380701
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.