Novus Biologicals
Manufacturer Code:NBP197761
Catalog # NBP197761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QRAESTATRPGPEK corresponding to internal region (near C-Terminus) according to NP_690872.2 NP_001093116.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Kidney-specific H(+)/organic cation antiporter; MATE-2; Multidrug and toxin extrusion protein 2; solute carrier family 47 (multidrug and toxin extrusion), member 2; Solute carrier family 47 member 2
Gene Aliases: MATE2; MATE2-B; MATE2-K; MATE2K; SLC47A2
UniProt ID: (Human) Q86VL8
Entrez Gene ID: (Human) 146802
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.