Invitrogen
Manufacturer Code:PA589905
Catalog # PIPA589905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 460-550 of human SLC47A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE-1; multidrug and toxin extrusion 1; Multidrug and toxin extrusion protein 1; rMATE-1; solute carrier family 47 (multidrug and toxin extrusion), member 1; Solute carrier family 47 member 1; solute carrier family 47, member 1
Gene Aliases: MATE1; RGD1311123; SLC47A1
UniProt ID: (Human) Q96FL8, (Rat) Q5I0E9
Entrez Gene ID: (Human) 55244, (Rat) 360539
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.