Invitrogen
Manufacturer Code:PA577554
Catalog # PIPA577554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)ESHGEIMMTDLEKK corresponding to amino acid residues 496-509 of mouse MATE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; MATE-1; MATE1; mMATE-1; Multidrug and toxin extrusion protein 1; rMATE-1; Solute carrier family 47 member 1; solute carrier family 47, member 1
Gene Aliases: 1300013J15Rik; AI036982; MATE1; mMATE1; RGD1311123; Slc47a1
UniProt ID: (Rat) Q5I0E9, (Mouse) Q8K0H1
Entrez Gene ID: (Rat) 360539, (Mouse) 67473
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.