Abnova
Manufacturer Code:PAB21466
Catalog # 89-122-961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC47A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC47A1.
For Research Use Only
Protein Aliases: FLJ10847; hMATE-1; MATE-1; MATE1; MGC64822; multidrug and toxin extrusion 1; Multidrug and toxin extrusion protein 1; solute carrier family 47 (multidrug and toxin extrusion), member 1; Solute carrier family 47 member 1; solute carrier family 47, member 1
Gene Aliases: MATE1; SLC47A1
UniProt ID: (Human) Q96FL8
Entrez Gene ID: (Human) 55244
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.