Novus Biologicals
Manufacturer Code:NBP189630
Catalog # NBP189630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AEEAALGPTEPAEGLSAPSLSPHCCPCRARLAFRNLGALLPRLHQLCCRMP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IPCA-2 IPCA-6 IPCA-8 PCANAP6IPCA6 PCANAP8 prostate cancer associated protein 2 prostate cancer associated protein 6 prostate cancer associated protein 8 prostate cancer-associated gene 2 prostate cancer-associated gene 6 prostate cancer-associated gene 8 Prostate cancer-associated protein 6 Prostein PRSTPCANAP2 solute carrier family 45 member 3 solute carrier family 45 member 3; prostate cancer associated protein 2; prostate cancer associated protein 6; prostate cancer associated protein 8; prostate cancer-associated gene 2; prostate cancer-associated gene 6; prostate cancer-associated gene 8; Prostate cancer-associated protein 6; Prostein; Solute carrier family 45 member 3; solute carrier family 45, member 3
Gene Aliases: IPCA-2; IPCA-6; IPCA-8; IPCA6; PCANAP2; PCANAP6; PCANAP8; PRST; SLC45A3
UniProt ID: (Human) Q96JT2
Entrez Gene ID: (Human) 85414
Molecular Function:
carbohydrate transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.