Invitrogen
Manufacturer Code:PA542515
Catalog # PIPA542515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC44A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Choline transporter-like protein 3; Solute carrier family 44 member 3; solute carrier family 44, member 3
Gene Aliases: BC010552; CTL3; RGD1305808; SLC44A3; UNQ558/PRO1115
UniProt ID: (Bovine) A5PK40, (Human) Q8N4M1, (Rat) Q6AY92, (Mouse) Q921V7
Entrez Gene ID: (Bovine) 507455, (Horse) 100058196, (Human) 126969, (Rabbit) 100350665, (Dog) 490152, (Rat) 295417, (Mouse) 213603, (Guinea pig) 100719379
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.